Learn More
Fisherbrand™ Colony Counter Bundle
Product Code. 17842966 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Display Type:
Digital
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Type Unit Size
17842966 Digital 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17842966
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
Estimated Shipment: 18-12-2026
Quickly identify and tally colonies with precision using this all-inclusive Colony Counter Bundle.
  • Quickly identify and tally colonies with precision using this all-inclusive package designed to streamline all colony counting procedures.
  • Fisherbrand eCount Colony Counter: Combines an electronic counter with a Sharpie™ pen for marking to prevent missing or double-counting colonies. Features count verification with beeps or LED flashes, dual counting modes (up and down), a display for total counts (up to 32 separate counts), and the ability to disable the counter for writing totals and notes.
  • Fisherbrand Lightbox: The Fisherbrand Lightbox serves as a LED light source for efficient bacterial colony counting. It features a spacious viewing area (125 x 100 mm) suitable for various samples, and is powered by six 1.5V AAA batteries (included).
  • Fisherbrand Magnifier: The Fisherbrand Mini-Magnifier enhances object visibility by providing 1.75X magnification. It features a swiveling, molded lens for comfortable observation.
  • Fisherbrand Silicone Lab Mat: This FDA food-grade silicone labmat is suitable for various laboratory settings, including cold rooms, clean rooms, and sterile suites, where high sterility is essential. Effectively safeguards countertops from temperatures up to 200°C (392°F) and demonstrates general chemical resistance to moderate oxidizing chemicals.
Specifications
Product Type Colony Counter with Lightbox, Magnifier and Lab Mat
Display Type Digital
Total Count Up to 999 colonies per zones, Up to 32 individual zones
Voltage 3 V