Learn More
Macherey-Nagel™ Collection Rack
Product Code. 10488981 Shop All Macherey-Nagel Products
Change view
Click to view available options
Volume (Metric):
96 x 1 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10488981 96 x 1 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10488981 Supplier Macherey-Nagel™ Supplier No. 738637
  • ONLINE EXCLUSIVE PRICE
    860.00 NOK
    790.66 NOK / Pack of 5
    Save 69.34 NOK
    8% Off
Estimated Shipment: 15-12-2026
Specifications
Type Collecting Rack
Volume (Metric) 96 x 1 mL
Includes Polypropylene Tube Strips
Quantity 5 Pack