Learn More
Macherey-Nagel™ CHROMABOND™ Multi 96 PP Reservoir Collection Vessels, 12 x 8-well Strips
Product Code. 10610162 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
10 Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10610162 10 Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10610162 Supplier Macherey-Nagel™ Supplier No. 738652
  • ONLINE EXCLUSIVE PRICE
    1070.00 NOK
    980.01 NOK / Pack of 10
    Save 89.99 NOK
    8% Off
Estimated Shipment: 09-12-2026
The Reservoir Collection Vessels are used as refills for the CHROMABOND™ Multi 96 Collection Rack for the Multi 96 Vacuum Manifold in solid phase extraction (SPE).
  • Accessory 12 x 8-well strip refills
  • For Multi 96 collection rack
  • Polypropylene
Specifications
Includes 12 x 8-well Roller Strips
Capacity (Metric) 1.0 mL (each tube)
For Use With (Application) Solid phase extraction vacuum manifold Multi 96
Material Polypropylene
Quantity 10 Pk.
Product Type Collecting Vessel