Learn More
STEMCELL Technologies Collagenase C, ACF
Product Code. p-7217478
Encompass_Preferred
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Product Code. 17109271
Brand: STEMCELL Technologies 07442
Animal component-free collagenase for the digestion of native collagen fibrils
  • Collagenase C, Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials
  • Collagenase is a protease consisting of a single polypeptide chain approximately 1,000 amino acid residues in length
  • Collagenase is capable of digesting native collagen fibrils commonly found in connective tissues and therefore is frequently used for tissue dissociation
  • Collagenase preparations contain the activity of several proteases including: collagenase, caseinase, clostripain, and trypsin (Kessler & Yaron)
  • Collagenase C, ACF contains low levels of tryptic activity, similar to Collagenase Type IV, and is intended for use in applications where it is necessary to prevent the introduction of potential animal-derived pathogens
Specifications
For Use With (Application) Digestion of Native Collagen Fibrils
Quantity 100 mg
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.