Learn More
STEMCELL Technologies Collagenase A, ACF
Description
- Collagenase A, Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials
- Collagenase is a protease consisting of a single polypeptide chain approximately 1,000 amino acid residues in length
- Collagenase is capable of digesting native collagen fibrils commonly found in connective tissues and therefore is frequently used for tissue dissociation
- Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler & Yaron)
- Collagenase A, ACF contains levels of proteolytic activity similar to type I and type II collagenases
- Collagenase A, ACF is intended for use in applications where it is necessary to prevent the introduction of potential animal-derived pathogens
Specifications
Specifications
| For Use With (Application) | Digestion of Native Collagen Fibrils |
| Quantity | 100 mg |