Learn More
STEMCELL Technologies Collagenase A, ACF
Product Code. 17169261
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Product Code. Quantity unitSize
17169261 100 mg 100mg
17179261 1 g 1g
2 options
Product Code. 17169261
Brand: STEMCELL Technologies 07434
Animal component-free collagenase for the digestion of native collagen fibrils
  • Collagenase A, Animal Component-Free (ACF) is obtained from cultures free of animal-derived materials
  • Collagenase is a protease consisting of a single polypeptide chain approximately 1,000 amino acid residues in length
  • Collagenase is capable of digesting native collagen fibrils commonly found in connective tissues and therefore is frequently used for tissue dissociation
  • Collagenase preparations contain the activity of several proteases, including collagenase, caseinase, clostripain, and trypsin (Kessler & Yaron)
  • Collagenase A, ACF contains levels of proteolytic activity similar to type I and type II collagenases
  • Collagenase A, ACF is intended for use in applications where it is necessary to prevent the introduction of potential animal-derived pathogens
Specifications
For Use With (Application) Digestion of Native Collagen Fibrils
Quantity 100 mg