Learn More
Colistin Sulfate Salt, MP Biomedicals
Product Code. 11460712 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
11460712 25 mg 25mg
1 options
Product Code. 11460712
Brand: MP Biomedicals 0219415725
Specifications
CAS 1264-72-8
Quantity 25 mg
compliance-icons
Product Identifier
  • Colistin Sulfate Salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Colistin Sulfate Salt