Learn More
Conductivity Solution, Oakton™
Product Code. 11870120 Shop All Cole-Parmer Products
Change view
Click to view available options
Conductivity:
0.023 mS (23 μS)
0.447 mS (447 μS)
1.413 mS (1413 μS)
1.5 mS (1500 μS)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Conductivity Unit Size
11870120 0.023 mS (23 μS) Each
11750049 0.447 mS (447 μS) Each
11754959 1.413 mS (1413 μS) Each
11765509 1.5 mS (1500 μS) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11870120 Supplier Cole Parmer Supplier No. 0065323
 Restricted Item
Estimated Shipment: 23-12-2026
Use this conductivity solution for accurate test results in the field or lab. Oakton™ Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
Use this conductivity solution for accurate test results in the field or lab. OaktonTM Conductivity Solutions have KCl, NaCl and 442 conversions printed on the bottle for ease of use.
  • Features temperature corrections printed on the bottle
  • Certified values listed in the tables and are referenced to 25°C
  • Certificates of analysis available separately
Specifications
Chemical Name or Material Cole-Parmer Conductivity Standards
Packaging Bottle
Conductivity 0.023 mS (23 μS)
Technique Calibration of Conductivity Instrument
Color Clear
pH 4.7 to 7.8
Boiling Point 100°C (212°F)
For Use With (Equipment) Conductivity Meter
Solubility Soluble
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL