Description
- Complies with FDA regulations governing direct food contact applications
- Available with straight or tapered blade
- Ends measure 7.9 x 50mm (WxL)
Specifications
Specifications
| Quantity | 2 Pk. |
| Autoclavable | Autoclavable |
| Shape | Half-rounded |
| Coating | Non-stick Fluo-Kem™ Teflon™ FEP Coating |
| Color | Silver/Brown |
| Certifications/Compliance | FDA |
| Product Type | Micro Spatula |
| Length (English) | 7.5 in. |
| Material | Stainless Steel |