Learn More
Lenz Laborglasintrumente™ Cold trap with Condensate Drain
Click to view available options
Volume (Metric):
200 mL
500 mL
Unit Size:
Each
Description
- With valve stopcock (with retaining device)
- For condensate drain
- Low cooling agent consumption due to double-walled design
- Apparatus connection: socket NS 29/32
- Vacuum pump connection: cone NS 29/32
- Made of DURAN™ tubing
Specifications
Specifications
| Material | DURAN Tubing |
| For Use With (Equipment) | Cooling Agents |
| Includes | Valve stopcock (with retaining device) for condensate drain |
| Quantity | 1 |
| Volume (Metric) | 500 mL |