Learn More
VACUUBRAND™ Cold Trap
Description
Cold trap SKF
Cold trap GKF
Inline oil filter HF
- Sturdy, easy to clean
- Easy to disassemble
- Two-wall design with good conductance
- Long operating time per coolant filling
- Easy condensate drainage without disassembling
Cold trap GKF
- Highly reflective insulation jacket for extended coolant life
- Vertical window: Direct observation of condensate and coolant levels
- PTFE stopcock: Drain condensate without disassembly
- Sheet metal shield for protection against external damage and implosion
Inline oil filter HF
- Extended oil lifetime
- Reduced service demands
- Easy space-saving mounting
- Simple filter change, service indicator
Specifications
Specifications
| Case Material | Stainless Steel, FKM, NBR |
| For Use With (Equipment) | Vacuubrand pumps |
| Type | SKF H 40 Cold Trap |