Learn More
VACUUBRAND™ Cold Trap
Artikelnummer. 10428181
Skift visning
Klik for at se tilgængelige muligheder
Type:
GKF 1000i Cold Trap
SKF H 25 Cold Trap
SKF H 40 Cold Trap
Pakningsstørrelse:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Type unitSize
10428181 SKF H 25 Cold Trap Each
10548381 GKF 1000i Cold Trap Each
10524902 SKF H 40 Cold Trap Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10428181 Leverandør VACUUBRAND™ Leverandørnr. 20667051
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Denne vare kan ikke returneres. Se returpolicy
Use this Cold Trap with very volatile solvents and reduce the back streaming of oil molecules into the vacuum system. Vacubrand™ Cold Traps provide important protection for your pump by separating condensates and aggressive media.
Cold trap SKF
  • Sturdy, easy to clean
  • Easy to disassemble
  • Two-wall design with good conductance
  • Long operating time per coolant filling
  • Easy condensate drainage without disassembling
Cold trap GKF
  • Highly reflective insulation jacket for extended coolant life
  • Vertical window: Direct observation of condensate and coolant levels
  • PTFE stopcock: Drain condensate without disassembly
  • Sheet metal shield for protection against external damage and implosion
Inline oil filter HF
  • Extended oil lifetime
  • Reduced service demands
  • Easy space-saving mounting
  • Simple filter change, service indicator
TRUSTED_SUSTAINABILITY
Tekniske data
Case Material Stainless Steel, FKM, NBR
For Use With (Equipment) Vacuubrand pumps
Type SKF H 25 Cold Trap
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.