Learn More
COIL Mouse anti-Human, Unconjugated, Clone: 2G12B8, Proteintech
Product Code. 16899735 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16899735 150 μL 150µL
16809745 20 μL 20µL
2 options
Product Code. 16899735
Brand: Proteintech 672181IG150UL
Mouse Monoclonal Antibody
The COIL antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human COIL. This antibody recognizes human antigen. The COIL antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Antigen COIL
Applications Western Blot, Immunofluorescence, ELISA
Classification Monoclonal
Clone 2G12B8
Concentration 500 μg/mL
Conjugate Unconjugated
Host Species Mouse
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 8161
Target Species Human
Isotype IgG1