Learn More
COD calibration standard 600mg/L NIST traceable, Reagecon™
Product Code. 11513162 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11513162 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11513162 Supplier Reagecon Supplier No. COD600
  • ONLINE EXCLUSIVE PRICE
    1610.00 NOK
    1499.83 NOK / 500mL
    Save 110.17 NOK
    6% Off
 Restricted Item
Estimated Shipment: 07-12-2026