missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COCO Rat anti-Mouse, Clone: 340531, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3356-SP
This item is not returnable.
View return policy
Description
COCO/DAND5 Monoclonal specifically detects COCO/DAND5 in Mouse samples. It is validated for Western Blot.Specifications
| COCO/DAND5 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q76LW6 | |
| DAND5 | |
| E. coli-derived recombinant mouse COCO Arg24-Leu185 Accession # Q76LW6 | |
| 25 μg | |
| Primary | |
| Detects mouse COCO in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) COCO, rhDAN, recombinant mouse (rm) Gremlin, rmCerberus, and recombinant chicken Caronte is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 340531 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CER2CRL2, Cerberus 2, Cerberus like-2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DAND5, Dante, DTE, FLJ38607, GREM3, GREM3cerberus 2, Gremlin-3, SP1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 199699 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction