Learn More
Cobimetinib, MedChemExpress
Product Code. 16491697
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16491697
Brand: Medchem Express HY13064
MedChemExpress Cobimetinib (GDC-0973, RG7420) is a potent, selective and oral MEK1 inhibitor with an IC50 of 4.2 nM for MEK1.
Cobimetinib (GDC-0973, RG7420) is a potent, selective and oral MEK1 inhibitor with an IC50 of 4.2 nM for MEK1.
Specifications
Chemical Name or Material Cobimetinib
CAS 934660-93-2
Color White
Purity Grade Notes Research
Molecular Formula C21H21F3IN3O2
Quantity 50 mg
Synonym GDC-0973 XL518
Solubility Information DMSO : ≥ 100 mg/mL (188.21 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES OC1([C@H]2NCCCC2)CN(C1)C(C3=C(C(F)=C(C=C3)F)NC4=C(C=C(C=C4)I)F)=O
Molecular Weight (g/mol) 531.31
Formula Weight 531.31
Percent Purity 98.79%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.