Learn More
3M™ Coarse Anti-Slip Tape
Description
- Anti-slip tape for light to heavy shoe-traffic areas
- High quality abrasive particles to ensure long-lasting friction in any application
- Highly durable surface withstands the rigors of equipment traffic
- Low-profile design helps reduce trip hazard
- High performance pressure-sensitive adhesive for easy application
- Applications: flat surfaces, steps, stairways, entrances, ramps, ladders, lawn equipment, snowmobiles, scooters, construction machinery and vehicles
- NFSI High Traction Certification
Specifications
Specifications
| Dimensions (L x W) | 15 x 30 m |
| Packaging Quantity | 1/Cs. |
| Color | Black |