Learn More
CNX-2006, MedChemExpress
Product Code. 16458537
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16458537
Brand: Medchem Express HY13897
MedChemExpress CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.
CNX-2006 is a mutant-selective and irreversible EGFR inhibitor with an IC50 below 20 nM for EGFRT790M.
Specifications
Chemical Name or Material CNX-2006
CAS 1375465-09-0
Color Gray
Purity Grade Notes Research
Molecular Formula C26H27F4N7O2
Quantity 5 mg
Solubility Information DMSO : ≥ 52 mg/mL (95.32 mM)
SMILES C=CC(NC1=CC=CC(NC2=NC(NC3=CC=C(NC4CN(CCF)C4)C=C3OC)=NC=C2C(F)(F)F)=C1)=O
Molecular Weight (g/mol) 545.53
Formula Weight 545.53
Percent Purity 99.68%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.