missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTROB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26880-1-AP-20UL
This item is not returnable.
View return policy
Description
CNTROB is a centrosomal protein that interacts with BRCA2 and is required for centriole duplication and cytokinesis (Zou et al., 2005 ).Specifications
| CNTROB | |
| Polyclonal | |
| Unconjugated | |
| CNTROB | |
| Centrobin, CNTROB, LIP8, LYST interacting protein 8, PP1221 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116840, 216846 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8CB62, Q8N137 | |
| Cntrob | |
| CNTROB Fusion Protein Ag25454 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction