Learn More
CNTROB Mouse anti-Human, Unconjugated, Clone: 3D12A4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67061-1-IG-150UL
Description
The CNTROB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CNTROB. This antibody recognizes human, mouse, rat, pig antigen. The CNTROB antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| CNTROB | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunofluorescence, ELISA | |
| 3D12A4 | |
| Unconjugated | |
| 150 μL | |
| 116840 | |
| IgG1 |