Learn More
CNTROB Mouse anti-Human, Unconjugated, Clone: 3D12A4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67061-1-IG-150UL
 View more versions of this product
Product Code. 16839465
  • 4836.10 NOK / 150µL
Description
The CNTROB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CNTROB. This antibody recognizes human, mouse, rat, pig antigen. The CNTROB antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
CNTROB
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunofluorescence, ELISA
3D12A4
Unconjugated
150 μL
116840
IgG1