missing translation for 'onlineSavingsMsg'
Learn More

CNTNAP3 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27410-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16895605

  • 3828.53 NOK / 150µL
Estimated Shipment: 12-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The specific function of this protein remains unknown. The protein encoded by this gene belongs to the NCP family of cell-recognition molecules. This family represents a distinct subgroup of the neurexins. NCP proteins mediate neuron-glial interactions in vertebrates and glial-glial contact in invertebrates. The protein encoded by this gene may play a role in cell recognition within the nervous system. Alternatively spliced transcript variants encoding different isoforms have been described but their biological nature has not been determined.
Specifications

Specifications

CNTNAP3
Polyclonal
Unconjugated
CNTNAP3
CASPR3, CNTNAP3, CNTNAP3A, KIAA1714, RP11 138L21.1, RP11 290L7.1
Rabbit
Antigen Affinity Chromatography
RUO
79937
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BZ76
CNTNAP3
CNTNAP3 Fusion Protein Ag26624
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers