missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13843-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Contactin 1 encodes a protein that is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
CNTN1 | |
Polyclonal | |
Unconjugated | |
CNTN1 | |
CNTN1, contactin 1, F3, Glycoprotein gp135, GP135, Neural cell surface protein F3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117258, 1272, 12805 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P12960, Q12860, Q63198 | |
Cntn1 | |
CNTN1 Fusion Protein Ag4803 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |