missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CNR1 Polyclonal antibody apecifically detetects CNR1 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
| Antigen | CNR1 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Concentration | 450 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide and 50% glycerol pH 7.3. |
| Gene Accession No. | BC074812 |
| Gene Symbols | CNR1 |
| Host Species | Rabbit |
| Immunogen | CNR1 fusion protein Ag31838 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?