missing translation for 'onlineSavingsMsg'
Learn More

CNR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 17669686 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17669686 20 μL 20µL
17659686 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17669686 Supplier Proteintech Supplier No. 297081AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CNR1 Polyclonal antibody apecifically detetects CNR1 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CNR1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 450 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. BC074812
Gene Symbols CNR1
Host Species Rabbit
Immunogen CNR1 fusion protein Ag31838
Purification Method Antigen affinity purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 1268
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.