Learn More
CNOT9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16851955 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16851955 20 μL 20µL
16841955 150 μL 150µL
2 options
Product Code. 16851955
Brand: Proteintech 225031AP20UL
Rabbit Polyclonal Antibody
This gene encodes a member of the highly conserved RCD1 protein family. The encoded protein is a transcriptional cofactor and a core protein of the CCR4-NOT complex. It may be involved in signal transduction as well as retinoic acid-regulated cell differentiation and development. Alternatively spliced transcript variants have been described for this gene.
Specifications
Antigen CNOT9
Applications Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CNOT9
Gene Accession No. Q5PQL2, Q92600, Q9JKY0
Gene Alias CNOT9, Rcd 1, RCD1, RCD1+, RQCD1
Gene Symbols Cnot9
Host Species Rabbit
Immunogen CNOT9 Fusion Protein Ag18197
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 301513, 58184, 9125
Target Species Human, Rat, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG