missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10752-1-AP-150UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against the full-length human CNOT8.
CNOT8 (CCR4-NOT transcription complex subunit 8), also known as CALIF or POP2, is a 292 amino acid protein that localizes to both the nucleus and the cytoplasm and functions as part of the CCR-NOT complex. Expressed ubiquitously, CNOT8 plays a role in transcriptional regulation for a diverse set of processes.Specifications
| CNOT8 | |
| Polyclonal | |
| Unconjugated | |
| CNOT8 | |
| CAF1, CAF1 like protein, CAF2, CALIF, CALIFp, CCR4 associated factor 8, CNOT8, hCAF1, POP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 363603, 69125, 9337 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9D8X5, Q9UFF9 | |
| Cnot8 | |
| CNOT8 Fusion Protein Ag1201 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering