missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNOT7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
3571.05 NOK - 5059.95 NOK
Specifications
| Antigen | CNOT7 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18261254
|
Novus Biologicals
NBP2-57301 |
100 μL |
5350.00 NOK 5059.95 NOK / 100µL Save 290.05 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668547
|
Novus Biologicals
NBP2-57301-25ul |
25 μL |
3780.00 NOK 3571.05 NOK / 25µL Save 208.95 NOK 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CNOT7 Polyclonal specifically detects CNOT7 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| CNOT7 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| BTG1 binding factor 1, BTG1-binding factor 1, CAF-1, CAF1carbon catabolite repressor protein (CCR4)-associative factor 1, CCR4-associated factor 1, CCR4-NOT transcription complex subunit 7, CCR4-NOT transcription complex, subunit 7, hCAF-1 | |
| CNOT7 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 29883 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:GHLYGLGSGSSYVQNGTGNAYEEEANKQS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title