Learn More
CNNM4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14066-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ancient conserved domain containing protein family. Members of this protein family contain a cyclin box motif and have structural similarity to the cyclins. The encoded protein may play a role in metal ion transport. Mutations in this gene are associated with Jalili syndrome which consists of cone-rod dystrophy and amelogenesis imperfecta.Specifications
CNNM4 | |
Polyclonal | |
Unconjugated | |
CNNM4 | |
ACDP4, CNNM4, cyclin M4, KIAA1592, Metal transporter CNNM4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26504, 363216, 94220 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P0C588, Q69ZF7, Q6P4Q7 | |
Cnnm4 | |
CNNM4 Fusion Protein Ag5121 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |