Description
- Ring-pull zip for easy donning and doffing when wearing gloves
- Double storm-facing stud closure for increased protection
- Helmet-compatible hood
- Raglan sleeve for greater freedom of movement
- Garment can be reused if not contaminated or damaged
- Stitched and high-frequency welded seams
- Tested to ASTM F903
Specifications
Specifications
| Color | Green |
| Hood Type | Attached |
| Material | PVC |
| Size | Medium |
| Ankle Style | Loose |
| Closure Type | Zipper |
| Cuff Style | Standard |
| Product Type | Chemical Protective Clothing |
| Seam Type | Welded |