Learn More
Emma Safety Footwear Clyde S3 Shoe
Product Code. 17512666
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
17512666 47 1 pair
17532656 39 1 pair
17542656 40 1 pair
17552656 41 1 pair
17562656 42 1 pair
17572656 43 1 pair
17582656 44 1 pair
17592656 45 1 pair
17502666 46 1 pair
17522666 48 1 pair
10 options
Product Code. 17512666
Brand: Emma Safety Footwear MM99554947
Only null left
Add to basket
Add to basket
If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clyde.
  • S3-Safetyshoe with sole plate for wet work
  • Metal Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clyde.
  • A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel. This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment
Specifications
Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black
For Use With (Application) Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Liner Material Hydro-Tec Antibacterial
Material Nubuck, Oiled Nubuck
Size 47
Fit Standard Width
Ankle Style High
Product Type Safety Shoe