missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CLTA (Human) Recombinant Protein

Human CLTA full-length ORF ( AAH19287, 1 a.a. - 218 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00001211-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123321

  • 3655.00 NOK / 10µg
Estimated Shipment: 16-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MAELDPFGAPAGAPGGPALGNGVAGAGEEDPAAAFLAQQESEIAGIENDEAFAILDGGAPGPQPHGEPPGGPDAVDGVMNGEYYQESNGPTDSYAAISQVDRLQSEPESIRKWREEQMERLEALDANSRKQEAEWKEKAIKELEEWYARQDEQLQKTKANNRAAEEAFVNDIDESSPGTEWERVARLCDFNPKSSKQAKDVSRMRSVLISLKQAPLVH
Specifications

Specifications

AAH19287
clathrin, light chain (Lca)
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CLTA
GST
1211
Wheat germ expression system
10 μg
LCA
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers