missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLPS Antibody (CLPS/9021), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24278AF647
This item is not returnable.
View return policy
Description
CLPS Monoclonal antibody specifically detects CLPS in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CLPS | |
| Monoclonal | |
| Alexa Fluor 647 | |
| colipase, colipase, pancreatic, pancreatic colipase preproprotein | |
| Recombinant fragment (around aa1-112) of human CLPS protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1208 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| CLPS/9021 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction