missing translation for 'onlineSavingsMsg'
Learn More

CLPP Mouse anti-Human, Porcine, Clone: 2E1D9, Proteintech

Product Code. p-7213074 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16847995 20 μL 20µL
16837995 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16847995

Brand: Proteintech 662711IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.

Specifications

Antigen CLPP
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2E1D9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CLPP
Gene Accession No. Q16740
Gene Alias CLPP, Endopeptidase Clp
Gene Symbols CLPP
Host Species Mouse
Immunogen CLPP Fusion Protein Ag8420
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100737271, 8192
Target Species Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.