Learn More
Clozapine
Product Code. p-7214487 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
16534264 1 g 1g
16544264 5 g 5g
2 options
Product Code. 16534264
Brand: Thermo Scientific Acros 464280010
Request Bulk or Custom Format
CAS: 5786-21-0 | C18H19ClN4 | 326.83 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5786-21-0
Molecular Formula C18H19ClN4
Molecular Weight (g/mol) 326.83
InChI Key ZUXABONWMNSFBN-UHFFFAOYSA-N
IUPAC Name 6-chloro-10-(4-methylpiperazin-1-yl)-2,9-diazatricyclo[9.4.0.0³,⁸]pentadeca-1,3,5,7,10,12,14-heptaene
SMILES CN1CCN(CC1)C1=C2C=CC=CC2=NC2=CC=C(Cl)C=C2N1
Specifications
Melting Point 182.0°C to 185.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: insoluble.
Formula Weight 326.82
Chemical Name or Material Clozapine
compliance-icons compliance-icons
Product Identifier
  • Clozapine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Clozapine