Learn More
Cloxacillin sodium salt, MP Biomedicals™
Product Code. 11462072 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462072 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11462072 Supplier MP Biomedicals Supplier No. 0215070205
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Type Cloxacillin Sodium Salt
Physical Form Crystalline Powder
CAS 7081-44-9
Color White
Quantity 5 g
Molecular Formula C19H19ClN3NaO6S
Synonym 5-methyl-3-(o-chlorophenyl)-4-isoxazolyl] pencillin
InChI Key KCUWTKOTPIUBRI-VICXVTCVSA-M
SMILES CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)[O-].O.[Na+]
IUPAC Name sodium;(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate;hydrate
Molecular Weight (g/mol) 475.876
PubChem CID 23675743
ChEBI CHEBI:34978
Formula Weight 457.9
Chemical Name or Material Cloxacillin sodium salt