Learn More
Cloquintocet-mexyl, TRC
Specifications
Specifications
| Chemical Name or Material | Cloquintocet-1-methylhexyl ester |
| CAS | 99607-70-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H22 Cl N O3 |
| Quantity | 1 g |
| Synonym | Acetic acid, 2-[(5-chloro-8-quinolinyl)oxy]-, 1-methylhexyl ester, Acetic acid, [(5-chloro-8-quinolinyl)oxy]-, 1-methylhexyl ester (9CI), 1-Methylhexyl 2-[(5-chloroquinolin-8-yl)oxy]acetate, CGA 185072, Cloquintocet 1-methylhexyl ester, Cloquintocet-mexyl |
| InChI Formula | InChI=1S/C18H22ClNO3/c1-3-4-5-7-13(2)23-17(21)12-22-16-10-9-15(19)14-8-6-11-20-18(14)16/h6,8-11,13H,3-5,7,12H2,1-2H3 |
| SMILES | CCCCCC(C)OC(=O)COc1ccc(Cl)c2cccnc12 |
| IUPAC Name | heptan-2-yl 2-(5-chloroquinolin-8-yl)oxyacetate |
| Molecular Weight (g/mol) | 335.83 |
| Show More |