Learn More
Cloquintocet-mexyl, Fluka™
Product Code. 16262862 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
16262862 100 mg 100mg
1 options
Product Code. 16262862
Brand: Honeywell-Fluka R1671100MG
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Analytical standard, reference material
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.
  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
  • Expert Technical Support
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cloquintocet-mexyl
Packaging Glass Bottle
CAS 99607-70-2
Color Beige
Melting Point 69.4°C
Boiling Point 275°C (Decomposes below the boiling point.)
Synonym rac-(2R)-heptan-2-yl [(5-chloroquinolin-8-yl)oxy]acetate
Industry Type Environmental Analysis
Molecular Weight (g/mol) 335.83
Quantity 100 mg
Grade Analytical Standard