Learn More
Clopidogrel Alcohol, Mikromol™
Description
Clopidogrel Alcohol
Specifications
Specifications
| Chemical Name or Material | Clopidogrel Alcohol |
| CAS | 1227294-01-0 |
| Analyte or Component Names | Clopidogrel Alcohol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H16 Cl N O S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Clopidogrel Alcohol |
| InChI Formula | InChI=1S/C15H16ClNOS/c16-13-4-2-1-3-12(13)14(10-18)17-7-5-15-11(9-17)6-8-19-15/h1-4,6,8,14,18H,5,7,9-10H2/t14-/m1/s1 |
| SMILES | OC[C@@H](N1CCc2sccc2C1)c3ccccc3Cl |
| IUPAC Name | (2S)-2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)ethanol |
| Show More |