missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CloneR™

Product Code. 17178301
Encompass
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 17178301

Brand: STEMCELL Technologies 05888

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Defined supplement for single-cell cloning of human ES and iPS cells

  • CloneR™ is a defined, serum-free supplement designed to increase the cloning efficiency and single-cell survival of human embryonic stem cells (ES cells) and induced pluripotent stem cells (iPS cells)
  • CloneR™ enables the robust generation of clonal cell lines without single-cell adaptation, thus minimizing the risk of acquiring genetic abnormalities
  • CloneR™ is compatible with the TeSR™ family of media for human ES and iPS cell maintenance as well as your choice of cell culture matrix

Specifications

Cell Type Pluripotent Stem Cells
For Use With (Application) Cell Culture
Quantity 10 mL
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.