Learn More
STEMCELL Technologies ClonaCell™-TCS Medium
Product Code. 17198201
Encompass
Change view
Click to view available options
Quantity:
80 mL
Unit Size:
80mL
Product Code. Quantity unitSize
17198201 80 mL 80mL
1 options
Product Code. 17198201
Brand: STEMCELL Technologies 03814
Semi-solid methylcellulose-based medium for selection and cloning of suspension-adapted and adherent cells (serum-containing)
  • ClonaCell™-TCS Medium is a serum-containing methylcellulose-based semi-solid medium that can be used for selecting and cloning a variety of suspension-adapted cell lines, including CHO-S and hybridomas
  • It can also be used for semi-solid cloning of some cell lines that grow adherently in the presence of serum, including CHO-K1, BHK-1, and HEK-293
  • This medium contains pre-selected fetal bovine serum (FBS) and bovine serum albumin (BSA) and supports robust growth of a wide variety of cell types
  • ClonaCell™-TCS Medium does not contain selection agents
Specifications
Cell Type Hybridomas; create dimCellLine - CHO Cells, HEK-293
For Use With (Application) Cell Culture; Semi-Solid Cloning
Quantity 80 mL