Learn More
STEMCELL Technologies ClonaCell™-CHO ACF Medium
Product Code. 17108211
Encompass
Change view
Click to view available options
Quantity:
90 mL
Unit Size:
90mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17108211 90 mL 90mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17108211 Supplier STEMCELL Technologies Supplier No. 03816
Serum-free, animal component-free semi-solid medium with supplements for selection and cloning of transfected CHO cells
  • ClonaCell™-CHO ACF is a methylcellulose-based semi-solid medium with supplements for the selection and cloning of suspension-adapted CHO cells in serum-free conditions
  • This medium is animal component-free and contains only recombinant proteins and synthetic components
  • Single-cell cloning efficiency of CHO cells is higher in this medium than in protein-free media, especially at low plating densities
  • It does not contain L-glutamine, selection agents, or phenol red and is compatible with dihydrofolate reductase (DHFR) and glutamine synthase (GS) selection systems
Specifications
Cell Line CHO
For Use With (Application) Cell Culture; Semi-Solid Cloning
Quantity 90 mL