Learn More
Clobetasol 17-Propionate, TRC
Product Code. 30134320
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134320 50 mg 50mg
30135239 1 g 1g
18002540 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134320 Supplier Toronto Research Chemicals Supplier No. TRCC58350050MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25122-46-7
Molecular Formula C25 H32 Cl F O5
Molecular Weight (g/mol) 466.97
Synonym Pregna-1,4-diene-3,20-dione, 21-chloro-9-fluoro-11β,17-dihydroxy-16β-methyl-, 17-propionate (8CI), (11β,16β)-21-Chloro-9-fluoro-11-hydroxy-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 21-Chloro-21-deoxybetamethasone 17-propionate, CCl 4725, CGP 9555, Clobederm, Clobesol, Clobetasol 17-propionate, Clobetasol propionate, Clobex, Dermoval, Dermovate, Dermoxin, Dermoxinale, GR 2/925, Karison, Olux, Olux-E, Skin Cap, Temovate, Tenovate
IUPAC Name [(8S,9R,10S,11S,13S,14S,16S,17R)-17-(2-chloroacetyl)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
SMILES CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CCl
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C25H32ClFO5/c1-5-21(31)32-25(20(30)13-26)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,27)19(29)12-23(18,25)4/h8-9,11,14,17-19,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1
Formula Weight 466.1922
Percent Purity >95%
Chemical Name or Material Clobetasol Propionate
Grade HPLC