missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLINT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10470-1-AP-150UL
This item is not returnable.
View return policy
Description
Binds to membranes enriched in phosphatidylinositol 4,5- bisphosphate (PtdIns(4,5)P2). May have a role in transport via clathrin-coated vesicles from the trans-Golgi network to endosomes. Stimulates clathrin assembly.Specifications
| CLINT1 | |
| Polyclonal | |
| Unconjugated | |
| CLINT1 | |
| clathrin interactor 1, CLINT, CLINT1, ENTH, Enthoprotin, EPN4, EPNR, Epsin 4, Epsin related protein, EPSINR, KIAA0171 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 216705, 360515, 9685 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14677, Q99KN9 | |
| Clint1 | |
| CLINT1 Fusion Protein Ag0754 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction