Learn More
Nickel Electro™ Clifton™ Bath Lab Armor™
Product Code. 12708898
Change view
Click to view available options
Capacity (Metric):
0.5 L
4 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12708898 4 L Each
12789008 0.5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12708898 Supplier Nickel Electro™ Supplier No. BA5
Only null left
Add to basket
Add to basket
Obtain superior water bath cleaning results when you replace water in your water bath with these thermally efficient pellets that are easy to keep clean. This Clifton™ Bath Armor™ tackles water contamination and protects your water bath from drying out.
  • Tackles water contamination by utilizing a dry, metallic thermal media comprised of small beads instead of water
  • Thermally efficient pellets that are easy to keep clean
  • Moisture and gas impermeable
  • High thermal conductivity
  • Smooth and rounded almost fluid like
  • Reduces water consumption
  • Saves the need for racks, floaties and bottle weights
  • Do not autoclave
Specifications
Capacity (Metric) 4 L
Type Bath Lab Armor
Quantity 4 L
Shape Round