Learn More
Thermo Scientific™ CLED Agar / MacConkey Agar No. 3
Description
Identify urinary tract pathogens clearly and accurately using Thermo Scientific™ CLED Agar / MacConkey Agar No. 3 combination bi-plates. CLED (Cysteine Lactose Electrolyte Deficient) Agar provides a valuable non-inhibitory diagnostic agar with good colonial differentiation and clear diagnostic characteristics. Detect the presence of Enterobacteriaceae with selective MacConkey Agar No. 3 while inhibiting Gram-positives.
The bi-plate combination of CLED Agar and MacConkey Agar No. 3 will help to specifically identify the relevant pathogens involved in urinary tract infections (UTI).
- Isolate, enumerate and presumptively identify common UTI related microorganisms
- Identify the degree of sample contamination
- Improved differentiation between coliforms and lactose and non-lactose fermenters
- Ready-to-use convenience of prepared media
Non-selective CLED Agar supports the growth of common urinary pathogens and provides good colonial differentiation with clear diagnostic characteristics. It is electrolyte deficient to prevent the swarming of Proteus spp. The presence of important contaminants, such as diphtheroids, lactobacilli and micrococci is also clearly elicited, giving an indication of the degree of contamination.
MacConkey Agar No. 3 is a highly selective medium suitable for detection and isolation of Enterobacteriaceae occurring in urinary tract infections. Due to the inclusion of a specially prepared fraction of bile salts, in addition to crystal violet, the medium gives improved differentiation between coliforms and non-lactose fermenting organisms while Gram-positive cocci are completely inhibited.
Not all products are available for sale in all territories.
Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2° to 12°C, in the dark |
| Description | CLED Agar / MacConkey Agar No. 3 |
| Format | 90mm biplate |
| Product Type | Agar Plate |
| Fill Weight | 17.5g ± 5% |
| Packaging Type | Wrapped in single cellulose-based film wrap |
| Quantity | 10/Pk. |