missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab CLED Agar
Click to view available options
:
CLED Agar; Form: Solid
Description
- Cystine, lactose, electrolyte deficient medium
- Recommended for the isolation and identification of urinary pathogenic bacteria.
Specifications
Specifications
| Form | Solid |
| Media Type | CLED Agar |
| Product Type | Agar |
| Formulation | Peptone 4.00 g/L, Tryptone 4.00 g/L, Meat Extract 3.00 g/L, Lactose 10.00 g/L, L-Cystine 0.128 g/L, Bromothymol Blue 0.020 g/L, Agar 15.00 g/L |
| For Use With (Application) | Isolation and Identification of Urinary Pathogenic Bacteria |
| Incubation Conditions | 37°C ± 1.0°C |
| Incubation Time | 21 ±3 hr. |
| pH | 7.4 ±0.2 |
| Quantity | 500 g |
| Solubility | Insoluble |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction