Learn More
CLEC4G Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18173-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a glycan-binding receptor and member of the C-type lectin family which plays a role in the T-cell immune response. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| CLEC4G | |
| Polyclonal | |
| Unconjugated | |
| CLEC4G | |
| CLEC4G, LP2698, LSECtin, UNQ431 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 339390 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6UXB4 | |
| CLEC4G | |
| CLEC4G Fusion Protein Ag12837 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |