missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLEC3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20311-1-AP-20UL
This item is not returnable.
View return policy
Description
Tetranectin (TN) is a serum and tissue protein, a C-type lectin, which binds to Ca++. It is a homotrimer of monomers each with a mass of 20 kDa, plasma or serum concentrations of TN are found to be approximately 10 mg/l. In vitro, TN can bind to kringle 4 of plasminogen and enhance the activation of plaminogen to plasmin, catalyzed by tissue plasminogen activator in the presence of poly-D-lysine. TN is best known as a prognostic marker in ovarian cancer.Specifications
| CLEC3B | |
| Polyclonal | |
| Unconjugated | |
| CLEC3B | |
| CLEC3B, Tetranectin, TN, TNA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21922, 7123 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05452, P43025 | |
| CLEC3B | |
| CLEC3B Fusion Protein Ag14147 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction