Learn More
Clebopride Malate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Clebopride Malate
Specifications
Specifications
| Chemical Name or Material | Clebopride malate |
| CAS | 57645-91-7 |
| Analyte or Component Names | Clebopride malate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 Cl N3 O2 . C4 H6 O5 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Clebopride malate, 4-Amino-N-(1-benzylpiperidin-4-yl)-5-chloro-2-methoxybenzamide acid (RS)-2-hydroxybutanedioate |
| InChI Formula | InChI=1S/C20H24ClN3O2.C4H6O5/c1-26-19-12-18(22)17(21)11-16(19)20(25)23-15-7-9-24(10-8-15)13-14-5-3-2-4-6-14;5-2(4(8)9)1-3(6)7/h2-6,11-12,15H,7-10,13,22H2,1H3,(H,23,25);2,5H,1H2,(H,6,7)(H,8,9) |
| SMILES | COc1cc(N)c(Cl)cc1C(=O)NC2CCN(Cc3ccccc3)CC2.OC(CC(=O)O)C(=O)O |
| IUPAC Name | 4-amino-N-(1-benzylpiperidin-4-yl)-5-chloro-2-methoxybenzamide;2-hydroxybutanedioic acid |
| Show More |