Learn More
Epredia™ ClearVue™ Mountants
Product Code. p-4532394 Shop All Epredia Products
Click to view available options
Solution Type:
Mountant, Toluene-based
Mountant, Xylene-based
Unit Size:
0.95 L
Each
Pack of 2
Product Code. 11376790
Brand: Epredia™ 4211
Enjoy compatibility with a variety of automated coverslippers to accommodate any lab setting with Epredia™ ClearVue™ Mountants.
TRUSTED_SUSTAINABILITY
Specifications
Color Light yellow
Product Type Mountant
Quantity 1 pt.
Solution Type Mountant, Toluene-based
Formulation Toluene, Acrylic Resin
Form Liquid
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Reproductive toxicity Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Flammable liquid Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.,May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.,Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.,Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P271-Use only outdoors or in a well-ventilated area.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.,In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P391-Collect spillage.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : toluene, acrylic resin