Learn More
Epredia™ ClearVue™ Coverslipper Coverslip Hoppers
Product Code. 12636376 Shop All Epredia Products
Change view
Click to view available options
Width (Metric):
40 mm
50 mm
55 mm
Length (Metric):
24 mm
Unit Size:
Pack of 1000
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Length (Metric) Unit Size
12636376 50 mm 24 mm Pack of 2
12688186 40 mm 24 mm Pack of 1000
13410497 55 mm 24 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12636376 Supplier Epredia™ Supplier No. A79210051
  • ONLINE EXCLUSIVE PRICE
    1530.00 NOK
    438.66 NOK / Pack of 2
    Save 1091.34 NOK
    71% Off
Estimated Shipment: 12-11-2026
Designed for use with the Epredia™ ClearVue™ coverlipper. ClearVue™ hoppers streamline coverslipping workflow.
  • Makes integration into the laboratory effortless
  • Ships preloaded with 500 or 575 slips each
  • Inserts quickly into the Epredia™ ClearVue™ coverslipper without workflow interruption
  • Requires no manual handling of glass
  • Dispenses a clean coverslip each and every time
TRUSTED_SUSTAINABILITY
Specifications
Product Type Coverslip Hopper
For Use With (Equipment) Epredia ClearVue Coverslipper
Quantity 2 x 500 Hoppers/Coverslips
Length (Metric) 24 mm
Thickness (Metric) 0.16 to 0.19 mm (# 1.5)
Width (Metric) 50 mm